AMBH303C5810 Ambeed, Inc.

Methyl 2-amino-5-bromo-3-iodobenzoate

  • MDL number MFCD00673015
  • Cas Number : 289039-83-4
  • Molecular Weight : 355.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/289039-83-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/289039-83-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart