AMBH303C5829 Ambeed, Inc.

N4,N4'-Di-p-tolyl-[1,1'-biphenyl]-4,4'-diamine

  • MDL number MFCD09833311
  • Cas Number : 10311-61-2
  • Molecular Weight : 364.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/10311-61-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/10311-61-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart