AMBH303C582D Ambeed, Inc.

Naquotinib

  • MDL number MFCD30496701
  • Cas Number : 1448232-80-1
  • Molecular Weight : 562.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (R)-5-((1-Acryloylpyrrolidin-3-yl)oxy)-6-ethyl-3-((4-(4-(4-methylpiperazin-1-yl)piperidin-1-yl)phenyl)amino)pyrazine-2-carboxamide
Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Normal
Country of Origin China
Product Link https://www.ambeed.com/products/1448232-80-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1448232-80-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart