AMBH303C5834 Ambeed, Inc.

1-Benzylpyrrolidine-3-carbonitrile oxalate

  • MDL number MFCD11501670
  • Cas Number : 1188265-21-5
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1188265-21-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1188265-21-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart