AMBH303C5837 Ambeed, Inc.

3-(Piperidin-4-ylmethyl)-1H-indole

  • MDL number MFCD01718591
  • Cas Number : 3515-49-9
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Flash Point 196.7
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 401.7�t 760 mmHg
Product Link https://www.ambeed.com/products/3515-49-9.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/3515-49-9.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart