AMBH303C5851 Ambeed, Inc.

Reserpine hydrochloride

  • MDL number MFCD23379918
  • Cas Number : 16994-56-2
  • Molecular Weight : 645.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1S,2R,3R,4aS,13bR,14aS)-methyl 2,11-dimethoxy-3-((3,4,5-trimethoxybenzoyl)oxy)-1,2,3,4,4a,5,7,8,13,13b,14,14a-dodecahydroindolo[2',3':3,4]pyrido[1,2-b]isoquinoline-1-carboxylate hydrochloride
Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/16994-56-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/16994-56-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart