AMBH303C5886 Ambeed, Inc.

1-(Isopropylamino)-3-(2-(2-methoxyethyl)phenoxy)propan-2-ol

  • MDL number MFCD18379554
  • Cas Number : 163685-38-9
  • Molecular Weight : 267.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/163685-38-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/163685-38-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart