AMBH303C5891 Ambeed, Inc.

Molsidomine

  • MDL number MFCD00869301
  • Cas Number : 25717-80-0
  • Molecular Weight : 242.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/25717-80-0.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312+P330-P501
MSDS https://www.ambeed.com/products/25717-80-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart