AMBH303C589B Ambeed, Inc.

Ibuprofen Lysine

  • MDL number MFCD01716155
  • Cas Number : 57469-77-9
  • Molecular Weight : 352.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-2,6-Diaminohexanoic acid compound with 2-(4-isobutylphenyl)propanoic acid (1:1)
Purity 95
Flash Point 216.7
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 319.6�t 760 mmHg
Product Link https://www.ambeed.com/products/57469-77-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/57469-77-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart