AMBH303C58EF Ambeed, Inc.

1-Aminopyrrole-2-carboxamide

  • MDL number MFCD11053506
  • Cas Number : 159326-69-9
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 425.9�t 760 mmHg
Product Link https://www.ambeed.com/products/159326-69-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/159326-69-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart