AMBH303C5914 Ambeed, Inc.

7-Bromo-1,3-dihydroimidazo[4,5-c]pyridin-2-one

  • MDL number MFCD03673684
  • Cas Number : 161836-12-0
  • Molecular Weight : 214.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 188.9�t 760 mmHg
Product Link https://www.ambeed.com/products/161836-12-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/161836-12-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart