AMBH303C591C Ambeed, Inc.

2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid dihydrochloride

  • MDL number MFCD19440796
  • Cas Number : 132210-25-4
  • Molecular Weight : 305.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/132210-25-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/132210-25-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart