AMBH303C5932 Ambeed, Inc.

Fimasartan

  • MDL number MFCD13194795
  • Cas Number : 247257-48-3
  • Molecular Weight : 501.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(1-((2'-(2H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-butyl-4-methyl-6-oxo-1,6-dihydropyrimidin-5-yl)-N,N-dimethylethanethioamide
Purity 98
Flash Point 372.9
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 693.0�0�t 760 mmHg
Product Link https://www.ambeed.com/products/247257-48-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/247257-48-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart