AMBH303C593A Ambeed, Inc.

1-(4-Chlorophenyl)-3-(4-methoxyphenyl)prop-2-en-1-one

  • MDL number MFCD00018708
  • Cas Number : 6552-63-2
  • Molecular Weight : 272.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/6552-63-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/6552-63-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart