AMBH303C5942 Ambeed, Inc.

5-Bromo-4-chloro-3-iodo-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD13193525
  • Cas Number : 1092579-75-3
  • Molecular Weight : 357.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1092579-75-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/1092579-75-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart