AMBH303C5951 Ambeed, Inc.

Isophthalic dihydrazide

  • MDL number MFCD00025117
  • Cas Number : 2760-98-7
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224°C
Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin China
Product Link https://www.ambeed.com/products/2760-98-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2760-98-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart