AMBH303C595B Ambeed, Inc.

Pentaerythritol tris(3-(1-aziridinyl)propionate)

  • MDL number MFCD01757346
  • Cas Number : 57116-45-7
  • Molecular Weight : 427.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(((3-(Aziridin-1-yl)propanoyl)oxy)methyl)-2-(hydroxymethyl)propane-1,3-diyl bis(3-(aziridin-1-yl)propanoate)
Purity 99
Flash Point 281.8
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 542.3�t 760 mmHg
Product Link https://www.ambeed.com/products/57116-45-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/57116-45-7.html
pictograms GHS07
hstatements H315-H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart