AMBH303C5963 Ambeed, Inc.

2-(Difluoromethoxy)-6-fluorobenzaldehyde

  • MDL number MFCD14698497
  • Cas Number : 1214333-68-2
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1214333-68-2.html

Safety Information

signalword Danger
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1214333-68-2.html
pictograms GHS02,GHS07
hstatements H226-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart