AMBH303C596B Ambeed, Inc.

Thiophene-2-sulfonic acid

  • MDL number MFCD11870146
  • Cas Number : 79-84-5
  • Molecular Weight : 164.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/79-84-5.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/79-84-5.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart