AMBH303C5984 Ambeed, Inc.

p-Tolylhydrazine hydrochloride

  • MDL number MFCD00012940
  • Cas Number : 637-60-5
  • Molecular Weight : 158.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/637-60-5.html

Safety Information

signalword Danger
pcodes P261-P301+P310-P305+P351+P338
MSDS https://www.ambeed.com/products/637-60-5.html
pictograms GHS06
hstatements H301-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart