AMBH303C5993 Ambeed, Inc.

5-Nitro-1H-inden-2(3H)-one

  • MDL number MFCD00598938
  • Cas Number : 116530-60-0
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132.5-135.5
Synonym N/A
Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 340.6�t 760 mmHg
Product Link https://www.ambeed.com/products/116530-60-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/116530-60-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart