AMBH303C59B0 Ambeed, Inc.

7,8-Dihydro-4H-thieno[3,2-b]azepin-5(6H)-one

  • MDL number MFCD00980799
  • Cas Number : 4751-61-5
  • Molecular Weight : 167.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/4751-61-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/4751-61-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart