AMBH303C59F3 Ambeed, Inc.

4-([1,1'-Biphenyl]-4-yl)-6-chloro-2-phenylpyrimidine

  • MDL number MFCD29477698
  • Cas Number : 1689538-58-6
  • Molecular Weight : 342.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1689538-58-6.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/1689538-58-6.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart