AMBH303C5A0F Ambeed, Inc.

N-(3,5-Dimethyladamantan-1-yl)formamide

  • MDL number MFCD01825787
  • Cas Number : 351329-88-9
  • Molecular Weight : 207.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69.5
Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 325�t 760 mmHg
Product Link https://www.ambeed.com/products/351329-88-9.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/351329-88-9.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart