AMBH303C5A3E Ambeed, Inc.

Methyl 4-hydroxy-2,5-dimethylbenzoate

  • MDL number MFCD13659356
  • Cas Number : 27023-05-8
  • Molecular Weight : 180.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/27023-05-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/27023-05-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart