AMBH303C5A5F Ambeed, Inc.

1-(Diethylamino)propan-2-ol

  • MDL number MFCD00004535
  • Cas Number : 4402-32-8
  • Molecular Weight : 131.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/4402-32-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/4402-32-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart