AMBH303C5A91 Ambeed, Inc.

Methyl 4-cyano-2-(trifluoromethyl)benzoate

  • MDL number MFCD28123953
  • Cas Number : 1641588-74-0
  • Molecular Weight : 229.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1641588-74-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1641588-74-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart