AMBH303C5AAB Ambeed, Inc.

3-(4-Chlorobenzylidene)isobenzofuran-1(3H)-one

  • MDL number MFCD02683417
  • Cas Number : 20526-97-0
  • Molecular Weight : 256.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/20526-97-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/20526-97-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart