AMBH303C5AD8 Ambeed, Inc.

2-(2-(Benzyloxy)-5-(tert-butyl)phenyl)-2-methylpropanenitrile

  • MDL number MFCD26393887
  • Cas Number : 1246213-27-3
  • Molecular Weight : 307.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1246213-27-3.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1246213-27-3.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart