AMBH303C5AD9 Ambeed, Inc.

4-(Di(1H-indol-3-yl)methyl)phenol

  • MDL number MFCD03723253
  • Cas Number : 151358-47-3
  • Molecular Weight : 338.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/151358-47-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/151358-47-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart