AMBH303C5AF1 Ambeed, Inc.

4-Methyl-3,4-dihydropyrrolo[1,2-a]pyrazin-1(2H)-one

  • MDL number MFCD14706806
  • Cas Number : 1243415-14-6
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1243415-14-6.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/1243415-14-6.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart