AMBH303C5AF4 Ambeed, Inc.

TCS-OX2-29

  • MDL number N/A
  • Cas Number : 372523-75-6
  • Molecular Weight : 397.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-1-(6,7-Dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)-3,3-dimethyl-2-((pyridin-4-ylmethyl)amino)butan-1-one
Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/372523-75-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/372523-75-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart