AMBH303C5AF5 Ambeed, Inc.

Methyl 4-chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylate

  • MDL number MFCD17016053
  • Cas Number : 1207518-63-5
  • Molecular Weight : 211.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1207518-63-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1207518-63-5.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart