AMBH303C5B04 Ambeed, Inc.

5,6-Dichloropyrazine-2,3-dicarboxylic acid

  • MDL number MFCD06208201
  • Cas Number : 59715-45-6
  • Molecular Weight : 237.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/59715-45-6.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/59715-45-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart