AMBH303C5B0B Ambeed, Inc.

6-(Chloromethyl)-N,N-dimethylpyridin-2-amine

  • MDL number MFCD10697582
  • Cas Number : 405103-62-0
  • Molecular Weight : 170.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/405103-62-0.html

Safety Information

signalword Danger
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/405103-62-0.html
pictograms GHS02,GHS07
hstatements H226-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart