AMBH303C5B53 Ambeed, Inc.

2-Chloro-3-(piperazin-1-yl)pyrazine hydrochloride

  • MDL number MFCD29761110
  • Cas Number : 66522-30-3
  • Molecular Weight : 235.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/66522-30-3.html

Safety Information

signalword Warning
pcodes P261-P280-P301+P312-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/66522-30-3.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart