AMBH303C5B71 Ambeed, Inc.

NS-6180

  • MDL number N/A
  • Cas Number : 353262-04-1
  • Molecular Weight : 323.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/353262-04-1.html

Safety Information

signalword Danger
pcodes P273-P280
MSDS https://www.ambeed.com/products/353262-04-1.html
pictograms GHS07,GHS09
hstatements H317-H411
Availability details for
Quantity
  • Availability
  • Add to Cart