AMBH303C5BC0 Ambeed, Inc.

2,6-Bis(1-methyl-1H-benzo[d]imidazol-2-yl)pyridin-4-ol

  • MDL number MFCD23135301
  • Cas Number : 533928-74-4
  • Molecular Weight : 355.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/533928-74-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/533928-74-4.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart