AMBH303C5BC4 Ambeed, Inc.

5-(Benzyloxy)pyrimidine-2-carbonitrile

  • MDL number MFCD09879704
  • Cas Number : 166672-22-6
  • Molecular Weight : 211.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 406.2�t 760 mmHg
Product Link https://www.ambeed.com/products/166672-22-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/166672-22-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart