AMBH303C5BCB Ambeed, Inc.

Phosphomolybdic acid hydrate

  • MDL number MFCD00149913
  • Cas Number : 51429-74-4
  • Molecular Weight : 259.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-71
Purity 72
Flash Point 129.2
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 320.2�0�t 760 mmHg
Product Link https://www.ambeed.com/products/51429-74-4.html

Safety Information

signalword Danger
pcodes P210-P220-P305+P351+P338-P310
hcodes 8,5.1
MSDS https://www.ambeed.com/products/51429-74-4.html
pictograms GHS03,GHS05
hstatements H272-H314
Availability details for
Quantity
  • Availability
  • Add to Cart