AMBH303C5BD3 Ambeed, Inc.

4-[(2,3-Epoxypropoxy)methyl]-2,2-dimethyl-1,3-dioxolane

  • MDL number MFCD07370127
  • Cas Number : 1607-37-0
  • Molecular Weight : 188.2200

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Physical Form liquid
Shipping Temperature Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1607-37-0.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/1607-37-0.html
pictograms GHS07
hstatements H227-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart