AMBH303C5BD5 Ambeed, Inc.

Phenylhydrazine hemisulfate

  • MDL number MFCD00070608
  • Cas Number : 52033-74-6
  • Molecular Weight : 314.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/52033-74-6.html

Safety Information

signalword Danger
pcodes P280-P301+P310-P305+P351+P338
MSDS https://www.ambeed.com/products/52033-74-6.html
pictograms GHS06
hstatements H301-H315-H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart