AMBH303C5BFB Ambeed, Inc.

2'-Chloro-2-fluoro-4'-methyl-[1,1'-biphenyl]-4-carboxylic acid

  • MDL number MFCD18320720
  • Cas Number : 1262008-90-1
  • Molecular Weight : 264.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1262008-90-1.html

Safety Information

signalword Warning
pcodes P233-P260-P261-P264-P270-P271-P280-P301+P312-P302+P352-P304-P304+P340-P305+P351+P338-P312-P321-P330-P332+P313-P337+P313-P340-P362-P403-P403+P233-P405-P501
MSDS https://www.ambeed.com/products/1262008-90-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart