AMBH303C5C37 Ambeed, Inc.

4-Bromo-7-chloroisoquinolin-1(2H)-one

  • MDL number MFCD13193317
  • Cas Number : 1028252-13-2
  • Molecular Weight : 258.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/1028252-13-2.html

Safety Information

signalword Warning
pcodes P261-P302+P352-P305+P351+P338
MSDS https://www.ambeed.com/products/1028252-13-2.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart