AMBH303C5C38 Ambeed, Inc.

6-Methylnaphthalen-2-ol

  • MDL number MFCD00046473
  • Cas Number : 17579-79-2
  • Molecular Weight : 158.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/17579-79-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/17579-79-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart