AMBH303C5C5F Ambeed, Inc.

PD123319 Ditrifluoroacetate

  • MDL number MFCD08276969
  • Cas Number : 136676-91-0
  • Molecular Weight : 736.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-1-(4-(Dimethylamino)-3-methylbenzyl)-5-(2,2-diphenylacetyl)-4,5,6,7-tetrahydro-1H-imidazo[4,5-c]pyridine-6-carboxylic acid compound with 2,2,2-trifluoroacetic acid (1:2)
Purity 98
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/136676-91-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/136676-91-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart