AMBH303C5C69 Ambeed, Inc.

4-Methyl-1H-benzo[d]imidazole-2(3H)-thione

  • MDL number MFCD03265647
  • Cas Number : 27231-33-0
  • Molecular Weight : 164.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/27231-33-0.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/27231-33-0.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart