AMBH303C5C88 Ambeed, Inc.

Copper(II) tetrafluoroborate hexahydrate

  • MDL number MFCD05863735
  • Cas Number : 72259-10-0
  • Molecular Weight : 345.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/72259-10-0.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/72259-10-0.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart