AMBH303C5C89 Ambeed, Inc.

Thiazolo[5,4-d]pyrimidin-2-amine

  • MDL number MFCD11847802
  • Cas Number : 920313-61-7
  • Molecular Weight : 152.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/920313-61-7.html

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/920313-61-7.html
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart