AMBH303C5CA5 Ambeed, Inc.

1,4-Bis(bromodifluoromethyl)benzene

  • MDL number MFCD02270920
  • Cas Number : 651-12-7
  • Molecular Weight : 335.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N/A
Purity 95
Storage temp. Sealed in dry,2-8
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Boiling point 235.5�t 760 mmHg
Product Link https://www.ambeed.com/products/651-12-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/651-12-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart